Pipette tips
Bevelled, graduated, 1 - 200 µl, -, 50 mm, natuur, Bulk, 1000 tips/bag
Catalogus # ALPCLW6391
Leverancier: Alpha Laboratories
Ga naar:
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Specificaties
- Kleurcode:natuur
- Verpakt:Bulk, 1000 tips/bag
- Steriel:-
- Volume:1 - 200 µl
- Lengte:50 mm
- Seq:07
- Tip stijl:Bevelled, graduated