51151401 PHOSPHORIC ACID 85% VLSI SELECT 1 * 2,5 L
Catalogus # 51151401.
Leverancier: BASF ELECTRONIC CHEMICALS
51151401 PHOSPHORIC ACID 85% VLSI SELECT 1 * 2,5 L
Catalogus # 51151401.
Leverancier: BASF ELECTRONIC CHEMICALS
Frequently Bought Together
Xyleen ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverancier: Merck
Ethanol, GPR RECTAPUR®
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
n-Hexaan, SupraSolv® for GC-ECD/FID, Supelco®
Leverancier: Merck
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
Leverancier: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Leverancier: Merck
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Leverancier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Xyleen ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Ethanol denatured, EMSURE® for analysis, Supelco®
Leverancier: Merck
Diethylether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Leverancier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
2-Propanol ≥99.8% VLSI, Semiconductor grade (Honeywell 17604)
Leverancier: Honeywell Chemicals