Container, with screw cap
1000 ml, 125 mm, 126 mm
Catalogus # HUEN915600
Leverancier: hünersdorff GmbH
Ga naar:
Product Details & Documents
Containers with screw cap round, for varied applications, made of food-safe natural polypropylene.
- Resistant to acids and chemicals
- With tightly closing lid
- Deep freezable
Specificaties
- Ø uitw.:125 mm
- Capaciteit:1000 ml
- Hoogte:126 mm
- Seq:03