Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Petrischalen

Petrischalen

With 3 vents, 60 mm, 8 mm, Steriel, 50×10/bag
Catalogus # ANIC05.80.18PADVI
Leverancier:   Anicrin
Petrischalen
Catalogus # ANIC05.80.18PADVI
Leverancier:  Anicrin
Petrischalen
Petrischalen

With 3 vents, 60 mm, 8 mm, Steriel, 50×10/bag

Price
254,00 €
Log in voor beschikbaarheid
1 * 1.000 ST
Specificaties
  • Ø:
    60 mm
  • Verpakt:
    50×10/bag
  • Steriliteit:
    Steriel
  • Uitvoering:
    With 3 vents
  • Hoogte:
    8 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.