Safety spectacles, Tracker
Antikras, antidamp, transparant, PC, kleurloos
Catalogus # 111-0390
Leverancier: Bollé Safety
Ga naar:
Product Details & Documents
These sporty design safety spectacles feature a durable PC frame, a smoke PC dual lens, straight temples with upper and lower protection and an adjustable headband.
- Interchangeable foam reinforcement with indirect ventilation
- Adjustable head strap and rubber arm tips provide a secure fit and greater protection
- ALS anti-fog/anti-scratch coating on both sides of clear and smoke lenses
- Rubberised eye seal provides exceptional protection against dust and other particles
Weight: 52 g
EN 166
Certificeringen: EN 166
Levering: Supplied with microfibre pouch.
Specificaties
- Type:Antikras, antidamp
- Kleur:transparant
- Seq:02
- Beglazing:PC, kleurloos