Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Gas regulators
Gas regulators
Two stage gas regualtor for ammonia/helium, brass body, PTFE seals, stainless steel diaphragms, 15 psig outlet pressure with 30 psig gauge, 1/8” swagelok outlet fitting, suits ICP-MS
Catalogus # HEWL5188-5374
Leverancier:   Agilent
Gas regulators
Catalogus # HEWL5188-5374
Leverancier:  Agilent
Gas regulators
Two stage gas regualtor for ammonia/helium, brass body, PTFE seals, stainless steel diaphragms, 15 psig outlet pressure with 30 psig gauge, 1/8” swagelok outlet fitting, suits ICP-MS
Price
4 730,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These gas regulators ensure that ICP-MS applications and methods are carried out with the consistent and stable gas flows. Effective gas regulation is vital for ensuring that ICP-MS systems operate from a stable basis allowing industry-leading levels of sensitivity to be achieved in relevant systems.
A range of specific models have been developed for various important gases and manufactured according to high standards.
Specificaties
  • Omschrijving:
    Two stage gas regualtor for ammonia/helium, brass body, PTFE seals, stainless steel diaphragms, 15 psig outlet pressure with 30 psig gauge, 1/8” swagelok outlet fitting, suits ICP-MS
  • Cat. No.:
    HEWL5188-5374
  • Max. inlaatdruk:
    207 bar