Orders LinkContactUsLinkComponent
Capillary and fitting kits
Capillary and fitting kits
1200 capillary kit for Int.Ø 0,12 mm
Catalogus # HEWLG1316-68716
Leverancier:   Agilent
Capillary and fitting kits
Catalogus # HEWLG1316-68716
Leverancier:  Agilent
Capillary and fitting kits
1200 capillary kit for Int.Ø 0,12 mm
Price
1 240,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These starter kits contain the most widely used capillary tubing, Swagelok connectors, and fittings in a variety of sizes.
  • Designed-in for highest chromatographic performance
  • High quality products for lowest instrument downtime and best total cost of ownership
  • OEM products fit tightest Agilent specifications for validated environments
We’ve also included our flexible stainless steel capillaries to help you make the best LC connections, regardless of equipment brand. Plus, as a special bonus, all multi-use kits (HEWL5065-9938, HEWL5065-9937, HEWL5065-9939) feature a FREE cybertool that puts more than 30 lab essentials at fingertips. For high-efficiency columns, it’s best to use narrow-diameter red tubing (Int.Ø 0,12 mm), instead of conventional green (Int.Ø 0,17 mm) tubing.
Specificaties
  • Omschrijving:
    1200 capillary kit for Int.Ø 0,12 mm
  • Seq:
    01
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.