Celtellers met drie rasters, McMaster
Cover glass with three counting grids (10×10 mm) divided into 10 parts
Catalogus # MARI0335001
Leverancier: MARIENFELD
Ga naar:
Product Details & Documents
This cell counter is suitable for liquid media.
- Three counting grids and spare cover glass
- Pair of clamps to fix the cover glass
- Ground edges and rounded corners
Specificaties
- Omschrijving:Cover glass with three counting grids (10×10 mm) divided into 10 parts