L(-)-Mannose ≥99%
Glass bottle for solids, 1 g
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Purity: 99% CAS No.: 10030-80-5 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 129-131° Beilstein Reference: 1724628
Synoniemen: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
- CAS-nummer:10030-80-5
- Formule:C₆H₁₂O₆
- MDL-nummer:MFCD00136021
- Molecular Weight:180.16 g/mol
- Kokend Pt:376 °C (1013 hPa)
- Smelten Pt:129…131 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:233-080-2
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:1 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



