MOPS-Na ≥98%
Glass bottle for solids, 25 g
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Purity: 98% CAS No.: 71119-22-7 Molecular Formula: C7H14NNaO4S Formula Weight: 231.25 Physical Form: Cryst powder Appearance: White Melting Point: ca 260° dec. MDL No.: MFCD00064350
Synoniemen: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
- CAS-nummer:71119-22-7
- Formule:C₇H₁₄NNaO₄S
- MDL-nummer:MFCD00064350
- Molecular Weight:231.25 g/mol
- Smelten Pt:∼ 260 °C (ontbinding)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:428-420-3
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:25 g



