Hexymethyleendiamine ≥98%
Glass bottle for solids, 500 g
Leverancier: Thermo Fisher Scientific
Hexymethyleendiamine ≥98%
Catalogus # A14212.36
Leverancier: Thermo Fisher Scientific
CAS-nummer: 124-09-4
Ga naar:
Product Details & Documents
Purity: 98+% CAS No.: 124-09-4 Molecular Formula: C6H16N2 Formula Weight: 116.21 Flash Point: 85°(185°F) Odour: Amine-like pH: 12.4 Melting Point: 39-43° Boiling Point: 199-204° Beilstein Reference: 1098307 Merck Reference: 14,4695 UN No.: 2280 MDL No.: MFCD00008243
Synoniemen: 1,6-Hexanediamine, Hexamethylenediamine
- CAS-nummer:124-09-4
- Formule:C₆H₁₆N₂
- MDL-nummer:MFCD00008243
- Molecular Weight:116.21 g/mol
- Merck index:14,4695
- Kokend Pt:199…204 °C
- Smelten Pt:39…43 °C
- Dichtheid:0,840
- Flits Pt:85 °C (185 °F)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:204-679-6
- UN:2280
- ADR:8,III
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:500 g