Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic
Bismuth(III) carbonate basic
Glass bottle for solids, 50 g
Catalog # A12483.18
Bismuth(III) carbonate basic
Catalog # A12483.18
CAS Number:  5892-10-4
Bismuth(III) carbonate basic
Bismuth(III) carbonate basic
Glass bottle for solids, 50 g
CAS Number:  5892-10-4
Price
35,70 €
Log in for availability
1 * 50 g
Product Details & Documents
CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877
Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:(BiO)₂CO₃
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Merck Index:14,1282
  • Density:6,86
  • Storage Temperature:Kamertemperatuur
  • EINECS:227-567-9
Specifications
Specifications
  • Verpakking:
    Glass bottle for solids
  • VPE:
    50 g
Specifications