Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic

Bismuth(III) carbonate basic

Glass bottle for solids, 1.000 g
Catalogus # A12483.0B
Leverancier:   Thermo Fisher Scientific
Bismuth(III) carbonate basic
Catalogus # A12483.0B
CAS-nummer:  5892-10-4
Bismuth(III) carbonate basic
Bismuth(III) carbonate basic

Glass bottle for solids, 1.000 g

CAS-nummer:  5892-10-4
Price
316,00 €
Log in voor beschikbaarheid
1 * 1.000 g
Product Details & Documents

CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877

Synoniemen:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS-nummer:5892-10-4
  • Formule:(BiO)₂CO₃
  • MDL-nummer:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Merck index:14,1282
  • Dichtheid:6,86
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:227-567-9
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    1.000 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.