Single channel pipettes, mechanical, fixed / variable volume, SoftGrip™
Product Details & Documents
Thanks to a soft contoured handle and a curved hilt these pipettes rest comfortable in your hand. The slim ejection sleeve is made from a high strength, chemically resistant polymer. The slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips.
- Built from high quality, state-of-the-art materials designed to provide years of reliable service
- All pipettes are completely autoclavable for easy decontamination and sterilisation without the need for disassembly and recalibration
- Isolated volume ring prevents accidental volume changes
- Magnified, easy-toread micrometer ensures reproducible, accurate volume setting
- Each pipette meets or exceeds industry standards for accuracy and precision when used with Hamilton Universal AdvanTip Precision pipette tips
Choose from six adjustable volume and ten fixed volume SoftGrip pipettes covering 0,2 µl to 1 ml. All pipettes have color-coded plunger buttons and labels for easy volume identification.
Services: Voor erkende kalibratie onder ISO8655 en ISO17025, onderhoud en reparatie mail naar [email protected]
Specificaties
- Kleurcode:rood
- Volume:30 - 300 µl
- Seq:05
- Stapgrootte:0,2 µl
- Juistheid:±1,2 (voor 30 µl); ±0,4 (voor 300 µl) %
- Onjuistheid:±0,4 (30 µl); ±0,15 (300 µl) %
- Aanbevolen tips:300 µl
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Pipetkalibratiesleutel, voor eenvoudige kalibratie in het veldPipetkalibratiesleutel, voor eenvoudige kalibratie in het veldHAMI34002Log in voor beschikbaarheid1 * 1 ST - 9,80 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Kalibratielabels voor pipetten
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Ejector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µlEjector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µlHAMI54756-01Log in voor beschikbaarheid1 * 1 ST - 23,80 €