Microplates, 96-well
F-bottom, clear, 40 - 340 µl, -, 382 µl
Catalogus # 738-0168
Leverancier: Greiner Bio-One
Ga naar:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Documents
Specificaties
- Aanbevolen werkvolume:40 - 340 µl
- Steriel:-
- Omschrijving:F-bottom, clear
- Seq:07
- Totaalvolume:382 µl
- Cat. No.:738-0168
- Milieu certificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lage impact op de fabricage