Basic fuchsin, Sigma-Aldrich®
Glass bottle for solids, 100 g
Catalog # 857343-100G
Supplier: SIGMA ALDRICH MICROSCOPY
Basic fuchsin, Sigma-Aldrich®
Catalog # 857343-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 632-99-5
Jump to:
Product Details & Documents
Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- CAS Number:632-99-5
- Formula:C₂₀H₂₀ClN₃
- MDL Number:MFCD00012569
- Molecular Weight:337.85 g/mol
- Merck Index:12,05686
- Melting Pt:235 °C
- Density:1,22 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Kamertemperatuur
- EINECS:211-189-6
Specifications
Specifications
- Verpakking:Glass bottle for solids
- VPE:100 g



