Labels, hazard warning
Hazard labels: laser beam, —, Polyester, zwart op geel, 100×87 mm, 3 labels/card
Catalogus # BRDY250228
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certificeringen: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Documents
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: laser beam
- Afmetingen:100×87 mm
- Materiaal:Polyester
- Seq:25
- Model:—
- Verpakt:3 labels/card



