Rubber Stoppers
45,0 mm, 50,5 mm, 59,5 mm
Catalogus # HECH41590053
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
DIN 12871, made of natural rubber, hardness 40 Shore A.
Documents
HECH41590041_TDS_EN
Technische fiche
Specificaties
- Ø onder:50,5 mm
- Ø boven:59,5 mm
- Hoogte:45,0 mm
- Seq:13