Rubber Stoppers
25,0 mm, 17,0 mm, 22,0 mm
Catalogus # HECH41590023
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
DIN 12871, made of natural rubber, hardness 40 Shore A.
Documents
HECH41590041_TDS_EN
Datasheet
Specificaties
- Ø onder:17,0 mm
- Ø boven:22,0 mm
- Hoogte:25,0 mm
- Seq:06