Formaldehyde 4%
20 L
Leverancier: KLINIPATH
Ga naar:
Product Details & Documents
Formaldehyde 4% is approximately equal to Formalin 10%
- pH 7
- Phosphate buffered
Synoniemen: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS-nummer:50-00-0
- Formule:CH₂O
- MDL-nummer:MFCD00003274
- Molecular Weight:30.03 g/mol
Documents
Specificaties
Specificatie testresultaten
- Formaldehyde (HCHO) g/l:38 - 42
- pH at 25 °C:6.8 - 7.2
- Sodium (Na):61 - 71 mmol/l
Specificaties
- VPE:20 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



