Jood ≥99.8%, solid ACS, Fluka™
Glass bottle, 500 g
Supplier: Honeywell Chemicals
Jood ≥99.8%, solid ACS, Fluka™
Catalog # 207772-500G
Supplier: Honeywell Chemicals
CAS Number: 7553-56-2
Jump to:
Product Details & Documents
Honeywell ACS solvents for general laboratory use are:
- A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested meet ACS general use specifications
- CAS Number:7553-56-2
- Formula:I₂
- MDL Number:MFCD00011355
- Molecular Weight:253.81 g/mol
- Melting Pt:113,5 °C
- Density:4,930 g/cm³ (20 °C)
- Storage Temperature:Kamertemperatuur
- EINECS:231-442-4
- UN:3495
- ADR:8,III
Specifications
Specification Test Results
- Assay:Min. 99.8 %
- Non-volatile matter:Max. 0.01 %
- Bromine and chlorine (as chloride):Max. 0.005 %
- Form:Solid
- Vapor Pressure:0.31 mmHg ( 25 °C)
- Resistivity:1.3E15 μΩ-cm
Specifications
- Verpakking:Glass bottle
- VPE:500 g
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



