Jood ≥99.8%, solid ACS, Fluka™
Glass bottle, 2,5 kg
Honeywell ACS solvents for general laboratory use are:
- A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested meet ACS general use specifications
- :7553-56-2
- :I₂
- :MFCD00011355
- :253.81 g/mol
- :113,5 °C
- :4,930 g/cm³ (20 °C)
- :Kamertemperatuur
- :231-442-4
- :3495
- :8,III
- Assay:Min. 99.8 %
- Non-volatile matter:Max. 0.01 %
- Bromine and chlorine (as chloride):Max. 0.005 %
- Form:Solid
- Vapor Pressure:0.31 mmHg ( 25 °C)
- Resistivity:1.3E15 μΩ-cm
- Verpakking:Glass bottle
- VPE:2,5 kg



