Orders LinkContactUsLinkComponent
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.
Primary solution red (1 ml solution contains 59,5 mg CoCl₂.6 H₂O) Reag. Ph. Eur., Glass bottle, 100 mL
Catalogus # REGEEPRS01
Leverancier:   REAGECON
Colour Reference Solutions, Reag.Ph.Eur.
Catalogus # REGEEPRS01
Leverancier:  REAGECON
CAS-nummer:  
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.
Primary solution red (1 ml solution contains 59,5 mg CoCl₂.6 H₂O) Reag. Ph. Eur., Glass bottle, 100 mL
CAS-nummer:  
Price
255,00 €
Log in voor beschikbaarheid
1 * 100 mL
Product Details & Documents
Specificaties
Specificaties
  • Verpakking:
    Glass bottle
  • VPE:
    100 mL
  • Omschrijving:
    Primary solution red (1 ml solution contains 59,5 mg CoCl₂.6 H₂O) Reag. Ph. Eur.