Nitrofen ≥98%
Glass bottle for solids, 25 g
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Purity: 98% CAS No.: 1836-75-5 Molecular Formula: C12H7Cl2NO3 Formula Weight: 284.10 Melting Point: 68-71° Beilstein Reference: 1887356 Merck Reference: 14,6598 UN No.: 3077 MDL No.: MFCD00128026
Synoniemen: 2,4-Dichloro-1-(4-nitrophenoxy)benzene
- CAS-nummer:1836-75-5
- Formule:C₁₂H₇Cl₂NO₃
- MDL-nummer:MFCD00128026
- Molecular Weight:284.1 g/mol
- Merck index:14,6598
- Smelten Pt:68…71 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:217-406-0
- ADR:9,III
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:25 g



