CombiPAL headspace syringes
23, 2500 µl, Fixed needle
Catalogus # HEWLG6500-80109
Leverancier: Agilent
Ga naar:
Product Details & Documents
Fixed needle headspace syringes feature a side port needle for gas flushing.
Micro seal is used to minimise instrument downtime and to prevent lost or compromised data caused by septum leaks and liner contamination.
Certifications: Conforms with strict CTC standards.
Specificaties
- Meter:23
- Type:Fixed needle
- Volume:2500 µl
- Seq:02