Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
CombiPAL headspace syringes
CombiPAL headspace syringes
23, 2500 µl, Fixed needle
Catalogus # HEWLG6500-80109
Leverancier:   Agilent
CombiPAL headspace syringes
Catalogus # HEWLG6500-80109
Leverancier:  Agilent
CombiPAL headspace syringes
CombiPAL headspace syringes
23, 2500 µl, Fixed needle
Price
429,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Fixed needle headspace syringes feature a side port needle for gas flushing.
Micro seal is used to minimise instrument downtime and to prevent lost or compromised data caused by septum leaks and liner contamination.
Certifications: Conforms with strict CTC standards.
Specificaties
  • Meter:
    23
  • Type:
    Fixed needle
  • Volume:
    2500 µl
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.