Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

pH bufferoplossingen, Pure Water™

pH buffer B, (475 ml bottle), 4,10, Plastic bottle, 4
Catalogus # 662-0977
Leverancier:   Thermo Orion
pH bufferoplossingen, Pure Water™
Catalogus # 662-0977
Leverancier:  Thermo Orion
CAS-nummer:  
pH bufferoplossingen, Pure Water™
pH bufferoplossingen, Pure Water™

pH buffer B, (475 ml bottle), 4,10, Plastic bottle, 4

CAS-nummer:  
Price
387,00 €
Log in voor beschikbaarheid
1 * 4 ST
Product Details & Documents

Buffers for calibration giving fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water, and process water.

  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
  • For best results, a high quality pH electrode is recommended

pHISA™Adjustor additive is designed to increase sample ionic strength without changing the pH of the sample.

Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    4
  • Omschrijving:
    pH buffer B, (475 ml bottle)
  • pH-waarde:
    4,10

Specificaties

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.