Slide Boxes
turkoois, PS, 50, 172×83×31 mm
Catalog # GLWAK50T
Supplier: GLW
Jump to:
Product Details & Documents
High quality slide storage boxes for microscopy slides.
Used for storing, transporting and freezing in science, industry and biotechnology.
Specifications
- Aantal objectglaasjes:50
- Kleur:turkoois
- Materiaal:PS
- Seq:06
- b×d×h:172×83×31 mm
Accessories
Product Information
- Catalog #OmschrijvingKleurMateriaalAvailabilityPrice
- GLWAZK20Divider for slides and coverslips of special sizesgrijsPSLog in for availability1 * 1 ST - 2,80 €
Product Information
- Catalog #OmschrijvingKleurMateriaalAvailabilityPrice
- GLWAZK15Divider for slides and coverslips of special sizeswitPSLog in for availability1 * 1 Un. - 5,94 €