Orders LinkContactUsLinkComponent
Mortars without pestles
Mortars without pestles
300 ml, 90 mm, 120 mm
Catalogus # BIBB718247
Leverancier:   SciLabware
Mortars without pestles
Catalogus # BIBB718247
Leverancier:  SciLabware
Mortars without pestles
Mortars without pestles
300 ml, 90 mm, 120 mm
Price
85,60 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Ideal for mixing and grinding powders.
  • Borosilicate glass
  • Low form model
Ordering information: Please order pestles separately.
Specificaties
  • Ø:
    120 mm
  • Capaciteit:
    300 ml
  • Hoogte:
    90 mm