[EN]MEPS-DEVELOPMENT KIT-CTC 23GA BINS P 1 * 5 ST
Catalog # 568-0065
Fournisseur: Trajan Scientific and Medical
[EN]MEPS-DEVELOPMENT KIT-CTC 23GA BINS P 1 * 5 ST
Catalog # 568-0065
Fournisseur: Trajan Scientific and Medical
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Sealing film for manual application, Parafilm® M
Fournisseur: HEATHROW
VWR®, gezichtstissues
Fournisseur: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
VWR®, Nitrilhandschoenen
Fournisseur: VWR Collection
pH-teststrips, MQuant®, Supelco®
Fournisseur: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschoenen
Fournisseur: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
Fournisseur: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
Fournisseur: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
Fournisseur: VWR Collection