Orders LinkContactUsLinkComponent
Flame atomisation systems
Flame atomisation systems
Mixing paddle, fluorinated
Catalogus # VARI110637000
Leverancier:   VARIAN
Flame atomisation systems
Catalogus # VARI110637000
Leverancier:  VARIAN
Flame atomisation systems
Flame atomisation systems
Mixing paddle, fluorinated
Price
55,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specificaties
  • Omschrijving:
    Mixing paddle, fluorinated
  • Seq:
    102
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    VARI6910009700
    Log in voor beschikbaarheid
    1 * 1 ST - 21,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Glass impact beads
    Glass impact beads
    VARI9910025700
    Log in voor beschikbaarheid
    1 * 5 ST - 192,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Aqueous spray chamber o-ring
    Aqueous spray chamber o-ring
    VARI9910093400
    Log in voor beschikbaarheid
    1 * 1 ST - 81,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Organics o-ring
    Organics o-ring
    VARI9910093500
    Log in voor beschikbaarheid
    1 * 1 ST - 1 100,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Fluorinated mixing paddles
    Fluorinated mixing paddles
    VARI9910093600
    Log in voor beschikbaarheid
    1 * 1 ST - 237,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Integrated liquid trap (fluorinated float and magnet)
    Integrated liquid trap (fluorinated float and magnet)
    VARI9910105800
    Log in voor beschikbaarheid
    1 * 1 ST - 401,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Nebuliser capillary kit
    Nebuliser capillary kit
    VARI9910093000
    Log in voor beschikbaarheid
    1 * 1 ST - 722,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Integrated nebuliser block
    Integrated nebuliser block
    VARI9910093100
    Log in voor beschikbaarheid
    1 * 1 ST - 1 280,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Flame atomisation system kit
    Flame atomisation system kit
    VARI190034100
    Log in voor beschikbaarheid
    1 * 1 ST - 2 720,00 €