Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Florisil® for chromatography

Florisil® for chromatography

Glass bottle for solids, 500 g
Catalogus # PROOSC-4179-B005
Leverancier:   LGC Standards PROMOCHEM
Florisil® for chromatography
Catalogus # PROOSC-4179-B005
CAS-nummer:  1343-88-0
Florisil® for chromatography

Glass bottle for solids, 500 g

CAS-nummer:  1343-88-0
Price
419,00 €
Log in voor beschikbaarheid
1 * 500 g
Product Details & Documents
Synoniemen:  Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
  • CAS-nummer:1343-88-0
  • Formule:MgO₃Si
  • MDL-nummer:MFCD00078249
  • Molecular Weight:100.38 g/mol
  • EINECS:215-681-1
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    500 g

Specificaties