Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fenofibrate, Reference Standards

Fenofibrate, Reference Standards

Fenofibrate Related Compound C, USP reference standard, Vial, 25 mg
Catalogus # 1269629.
Leverancier:   USP
Fenofibrate, Reference Standards
Catalogus # 1269629.
Leverancier:  USP
CAS-nummer:  
Fenofibrate, Reference Standards

Fenofibrate Related Compound C, USP reference standard, Vial, 25 mg

CAS-nummer:  
Price
1 240,00 €
Log in voor beschikbaarheid
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Vial
  • VPE:
    25 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Fenofibrate Related Compound C
  • Cat. No.:
    1269629.
  • Omschrijving:
    Fenofibrate Related Compound C
  • Formule:
    C₂₄H₂₇ClO₆
  • Synonym(s):
    1-methylethyl 2-[[2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoyl]oxy]-2-methylpropanoate
  • CAS No.:
    217636-48-1

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.