Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Barium 99+%, rods, Ø ∼ 2 cm

Barium 99+%, rods, Ø ∼ 2 cm

Plastic bottle, 100 g
Catalogus # ACRO317861000
Ontvlambaar
Gezondheid
Leverancier:   Thermo Fisher Scientific
Barium 99+%, rods, Ø ∼ 2 cm
Catalogus # ACRO317861000
CAS-nummer:  7440-39-3
Barium 99+%, rods, Ø ∼ 2 cm
Barium 99+%, rods, Ø ∼ 2 cm

Plastic bottle, 100 g

CAS-nummer:  7440-39-3
Price
223,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
  • CAS-nummer:7440-39-3
  • Formule:Ba
  • MDL-nummer:MFCD00134031
  • Molecular Weight:137.33 g/mol
  • Merck index:12,00991
  • Kokend Pt:1640 °C
  • Smelten Pt:725 °C
  • Dichtheid:3,51 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:231-149-1
  • ADR:4.3,II
  •  :Density: 3,51 g/cm³ (20 °C)
  • UN:1400

Documents

Specificaties

Specificatie testresultaten

  • Appearance:
    Grey Rods
  • Assay:
    ≥99.0 % Ba (incl. Sr)
  • Strontium (Sr):
    ≤0.8 %

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    100 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.