Methyleenchloride 99+% stabilised, extra pure
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Methyleenchloride 99+% stabilised, extra pure
Catalog # ACRO383780025
Supplier: Thermo Fisher Scientific
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39…40 °C (760 mmHg)
- Melting Pt:–97 °C
- Density:1,325 g/cm³
- Storage Temperature:Kamertemperatuur
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with amylene
- UN:1593
Documents
ACRO38378
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (K.F.)
- GC:≥99.0 %
- Stabilizer:Approx. 100 ppm Amylene
- Residue after evaporation:≤0.05 %
Specifications
- Verpakking:Glass bottle
- VPE:2,5 L
- Cat. No.:ACRO383780025
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



