Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Methyleenchloride 99+% stabilised, extra pure

Methyleenchloride 99+% stabilised, extra pure

Metal drum, 10 L
Catalogus # ACRO124050100
Gezondheid
Irriterend
Leverancier:   Thermo Fisher Scientific
Methyleenchloride 99+% stabilised, extra pure
Catalogus # ACRO124050100
CAS-nummer:  75-09-2
Methyleenchloride 99+% stabilised, extra pure
Methyleenchloride 99+% stabilised, extra pure

Metal drum, 10 L

CAS-nummer:  75-09-2
Price
286,00 €
Log in voor beschikbaarheid
1 * 10 L
Product Details & Documents
Synoniemen:  Methylene chloride, DCM
  • CAS-nummer:75-09-2
  • Formule:CH₂Cl₂
  • MDL-nummer:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck index:12,06140
  • Kokend Pt:39,8 °C (1013 hPa)
  • Smelten Pt:–97 °C
  • Dichtheid:1,325 g/cm³
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilised with ethanol
  • UN:1593

Documents

Specificaties

Specificatie testresultaten

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥99.0 %
  • Stabilizer:
    Approx. 0.2 % Ethanol
  • Residue after evaporation:
    ≤0.05 %

Specificaties

  • Verpakking:
    Metal drum
  • VPE:
    10 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.