Methyleenchloride 99+% stabilised, extra pure
Plastic bottle, 5 L
Leverancier: Thermo Fisher Scientific
Methyleenchloride 99+% stabilised, extra pure
Catalogus # ACRO124050050
Leverancier: Thermo Fisher Scientific
CAS-nummer: 75-09-2
Ga naar:
Product Details & Documents
Synoniemen: Methylene chloride, DCM
- CAS-nummer:75-09-2
- Formule:CH₂Cl₂
- MDL-nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck index:12,06140
- Kokend Pt:39,8 °C (1013 hPa)
- Smelten Pt:–97 °C
- Dichtheid:1,325 g/cm³
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with ethanol
- UN:1593
Specificaties
Specificatie testresultaten
- Appearance:Clear colorless Liquid
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Stabilizer:Approx. 0.2 % Ethanol
- Residue after evaporation:≤0.05 %
Specificaties
- Verpakking:Plastic bottle
- VPE:5 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



