Methyleenchloride 99.8+% stabilised for analysis
Metal drum, 10 L
Leverancier: Thermo Fisher Scientific
Methyleenchloride 99.8+% stabilised for analysis
Catalogus # 113460100.
Leverancier: Thermo Fisher Scientific
CAS-nummer: 75-09-2
Ga naar:
Product Details & Documents
Synoniemen: Methylene chloride, DCM
- CAS-nummer:75-09-2
- Formule:CH₂Cl₂
- MDL-nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck index:12,06140
- Kokend Pt:39,8 °C (1013 hPa)
- Smelten Pt:–97 °C
- Dichtheid:1,325 g/cm³
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with amylene
- UN:1593
Documents
ACRO11346
SDS
Specificaties
Specificatie testresultaten
- Appearance:Clear liquid
- Water:≤0.02 % (Coulometric)
- GC:≥99.80 %
- Color scale:≤10 APHA
- Stabilizer:15 to 100 ppm Amylene
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0003 meq/g
- Substance darkened by H2SO4:≤5 APHA
- Free chlorine:≤1 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
Specificaties
- Verpakking:Metal drum
- VPE:10 L
- Cat. No.:113460100.



