Orders LinkContactUsLinkComponent
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffers I-VI trial kit (includes each 10 ml), 1 KIT
Catalogus # 786-234
Leverancier:   G-Biosciences
Extraction buffers, FOCUS™
Catalogus # 786-234
Leverancier:  G-Biosciences
CAS-nummer:  
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffers I-VI trial kit (includes each 10 ml), 1 KIT
CAS-nummer:  
Price
372,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
FOCUS™ Extraction Buffers are experimentally optimised concentrations of critical agents, buffering and stabilising agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulphobetaines (SB). They are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    FOCUS™ Extraction Buffers I-VI trial kit (includes each 10 ml)
Specificaties