G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl), 1 L
Fournisseur: G-Biosciences
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
N° de catalogue R017
Fournisseur: G-Biosciences
Numéro CAS:
Aller à :
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
1 M Tris·HCl (pH 7,4)
- 1,5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- VPE:1 L
- Omschrijving:Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)