Orders LinkContactUsLinkComponent
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH), 1 gallon
Catalogus # R016
Leverancier:   G-Biosciences
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Catalogus # R016
Leverancier:  G-Biosciences
CAS-nummer:  
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH), 1 gallon
CAS-nummer:  
Price
125,00 €
Log in voor beschikbaarheid
1 * 4 L
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1.5 M sodium chloride
  • 0.5 M  sodium hydroxide
A high-pH reagent used to denature double-stranded nucleic acids into single strands, preparing DNA or RNA for hybridization. Ready-to-use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
Specificaties
Specificaties
  • VPE:
    1 gallon
  • Omschrijving:
    Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH)
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.