Orders LinkContactUsLinkComponent
DNAOUT™ DNA Removal Solution
DNA OUT™, spray bottle, 250 mL
Catalogus # 786-74
Leverancier:   G-Biosciences
DNAOUT™ DNA Removal Solution
Catalogus # 786-74
Leverancier:  G-Biosciences
CAS-nummer:  
DNAOUT™ DNA Removal Solution
DNAOUT™ DNA Removal Solution
DNA OUT™, spray bottle, 250 mL
CAS-nummer:  
Price
58,40 €
Log in voor beschikbaarheid
1 * 250 mL
Product Details & Documents
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
  • Ready-to-use with convenient spray bottle and refill bottles available
  • Remove contaminating DNA prior to sensitive work, including PCR
  • Removes amplicon, plasmid and genomic DNA
  • Fast acting (10-15 seconds)
  • Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more
Specificaties
Specificaties
  • VPE:
    250 mL
  • Omschrijving:
    DNA OUT™, spray bottle
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.