Orders LinkContactUsLinkComponent
RNaseOUT™ RNAse removal solution
RNaseOUT™ RNAse removal solution
RNaseOut™, spray bottle, 250 mL
Catalogus # 786-70
Leverancier:   G-Biosciences
RNaseOUT™ RNAse removal solution
Catalogus # 786-70
Leverancier:  G-Biosciences
CAS-nummer:  
RNaseOUT™ RNAse removal solution
RNaseOUT™ RNAse removal solution
RNaseOut™, spray bottle, 250 mL
CAS-nummer:  
Price
56,10 €
Log in voor beschikbaarheid
1 * 250 mL
Product Details & Documents
RNaseOUT™ is uniquely formulated to destroy and remove RNases on contact.
  • Simply spray and rinse
  • RNaseOUT™ is non-toxic and residue free
  • RNaseOUT™ allows common equipment to be safely and confidently used for all RNA work
Specificaties
Specificaties
  • VPE:
    250 mL
  • Omschrijving:
    RNaseOut™, spray bottle