Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Iodoacetyl-LC-biotin, 2 mg, 8
786-085
The OneQuant™ format prevents loss of reagent due to repeated weighing as each vial contains only 1 - 2 mg HOOK™ Biotin Reagent.
- VPE:8
- Omschrijving:OneQuant™ HOOK™ Iodoacetyl-LC-biotin, 2 mg