Orders LinkContactUsLinkComponent
Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Sulfo-NHS-LC-biotin, 1 mg, 8
  786-084
Biotin reagents, OneQuant™ HOOK™
  786-084
 :  
Biotin reagents, OneQuant™ HOOK™
Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Sulfo-NHS-LC-biotin, 1 mg, 8
 :  
260,00 €
1 * 8 ST
The OneQuant™ format prevents loss of reagent due to repeated weighing as each vial contains only 1 - 2 mg HOOK™ Biotin Reagent.
  • VPE:
    8
  • Omschrijving:
    OneQuant™ HOOK™ Sulfo-NHS-LC-biotin, 1 mg