Natriumhydroxide 25% in aqueous solution, J.T.Baker®
Plastic bottle, 5 L
Leverancier: Avantor
Natriumhydroxide 25% in aqueous solution, J.T.Baker®
Catalogus # 2613.5000PE
Leverancier: Avantor
CAS-nummer: 1310-73-2
Ga naar:
Product Details & Documents
Synoniemen: Caustic soda
- CAS-nummer:1310-73-2
- Formule:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Kokend Pt:119…135 °C (1013 hPa)
- Smelten Pt:9…10 °C
- Dichtheid:1,53 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Specificaties
Specificatie testresultaten
- Assay (NaOH):24.4 - 25.6 %
- Appearance (clear, colorless liquid):passes test
- Endotoxin Concentration (EU/ml):max. 2.0
- Sodium Carbonate (Na2CO3):max. 0.6 %
- Trace Impurities (in ppm)Chlorides (as Cl):max. 5
- Trace Impurities (in ppm)Iron (Fe):max. 0.5
- Trace Impurities (in ppm)Heavy Metals (as Pb):max. 1
Specificaties
- Verpakking:Plastic bottle
- VPE:5 L



