Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Petrischalen

Petrischalen

Contact plate, with 3 vents, 90 mm, 15 mm, Steriel, 25×20/bag
Catalogus # ANIC05.10.18PAKI
Leverancier:   Anicrin
Petrischalen
Catalogus # ANIC05.10.18PAKI
Leverancier:  Anicrin
Petrischalen
Petrischalen

Contact plate, with 3 vents, 90 mm, 15 mm, Steriel, 25×20/bag

Price
206,00 €
Log in voor beschikbaarheid
1 * 500 ST
Product Details & Documents
Specificaties
  • Ø:
    90 mm
  • Verpakt:
    25×20/bag
  • Steriliteit:
    Steriel
  • Uitvoering:
    Contact plate, with 3 vents
  • Hoogte:
    15 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.