Orders LinkContactUsLinkComponent
Adapters, delivery, straight
24/29 NS, 100 mm
Catalogus # WITG1.900.024
Leverancier:   witeg Labortechnik
Adapters, delivery, straight
Catalogus # WITG1.900.024
Leverancier:  witeg Labortechnik
Adapters, delivery, straight
Adapters, delivery, straight
24/29 NS, 100 mm
Price
23,10 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specificaties
  • Huls:
    24/29 NS
  • Buislengte:
    100 mm