Orders LinkContactUsLinkComponent
Objectives for CKX53
Objectives for CKX53
Plan achromatic fluorine objective, Ph 4×/0,13, 17 mm
Catalog # OLYMN1479100
Objectives for CKX53
Catalog # OLYMN1479100
Supplier:  
Objectives for CKX53
Objectives for CKX53
Plan achromatic fluorine objective, Ph 4×/0,13, 17 mm
Discontinued
Product Details & Documents
Specifications
  • Vergroting:
    Ph 4×/0,13
  • Omschrijving:
    Plan achromatic fluorine objective
  • Seq:
    004
  • Werkafstand:
    17 mm