Objectives for CKX53
LC achromatic objective with 1 mm correction and relief contrast (RC3), 40×/0,55, 1,9 mm
Catalogus # OLYMN1478100
Ga naar:
Discontinued
Product Details & Documents
Specificaties
- Vergroting:40×/0,55
- Omschrijving:LC achromatic objective with 1 mm correction and relief contrast (RC3)
- Werkafstand:1,9 mm