Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
45/40, 65 mm
Catalogus # COWI010.1245
Leverancier:   COWIE
Stoppers, self realasing, PTFE
Catalogus # COWI010.1245
Leverancier:  COWIE
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
45/40, 65 mm
Price
60,20 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specificaties
  • (NS):
    45/40
  • Hoogte:
    65 mm